Anti-CDCP1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 12K58]

Lieferant: United States Biological
USBIC2566-01KEA 0 EUR
USBIC2566-01K
Anti-CDCP1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 12K58]
Antikörper
Do not freeze!

Type: Primary
Antigen: CDCP1 (CUB domain containing protein 1)
Clonality: Monoclonal
Clone: 12K58
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR