Anti-CDC25C Mouse Monoclonal Antibody [clone: 1F12]

Lieferant: United States Biological
USBIC2560-05NEA 633 EUR
USBIC2560-05N
Anti-CDC25C Mouse Monoclonal Antibody [clone: 1F12]
Antikörper
Recommended dilution: Western Blot: 1:500-1:2000; Immunochemistry: 1:200

Type: Primary
Antigen: CDC25C (cell division cycle 25C)
Clonality: Monoclonal
Clone: 1F12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR