Anti-CDC25A Rabbit Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBIC2560-01Z-BIOTEA 678 EUR
USBIC2560-01Z-BIOT
Anti-CDC25A Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: CDC25A (cell division cycle 25A)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR