Anti-CDC6 Mouse Monoclonal Antibody [clone: 10K145]

Lieferant: United States Biological
USBIC2550-05CEA 440 EUR
USBIC2550-05C
Anti-CDC6 Mouse Monoclonal Antibody [clone: 10K145]
Antikörper
Recommended dilution: Western Blot: 1-2?g/ml

Type: Primary
Antigen: CDC6 (cell division cycle 6)
Clonality: Monoclonal
Clone: 10K145
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR