Anti-CDK1 Mouse monoclonal antibody APC (Allophycocyanin) [clone: 10B2173]

USBIC2549-49N-APCEA 0 EUR
USBIC2549-49N-APC
Anti-CDK1 Mouse monoclonal antibody APC (Allophycocyanin) [clone: 10B2173]
Type: Primary
Antigen: CDK1 (cyclin-dependent kinase 1)
Clonality: Monoclonal
Clone: 10B2173
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR