Anti-LAIR1 Goat Polyclonal Antibody

Lieferant: United States Biological
USBIC2549-32CEA 471 EUR
USBIC2549-32C
Anti-LAIR1 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1-3?g/ml

Type: Primary
Antigen: LAIR1 (leukocyte-associated immunoglobulin-like receptor 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR