Anti-CD302 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Lieferant: United States Biological
USBIC2549-30-FITCEA 0 EUR
USBIC2549-30-FITC
Anti-CD302 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: CD302 (CD302 molecule)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR