Anti-CD274 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 9E32]
Lieferant: United States Biological
USBIC2549-22F4EA 511 EUR
USBIC2549-22F4
Anti-CD274 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 9E32]
Antikörper
Type: Primary
Antigen: CD274 (CD274 molecule)
Clonality: Monoclonal
Clone: 9E+32
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse