Anti-CD274 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIC2549-22CEA 493 EUR
USBIC2549-22C
Anti-CD274 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.5-1 ?g/ml; Immunochemistry: 2.5 ?g/ml

Type: Primary
Antigen: CD274 (CD274 molecule)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR