Anti-CD270 Mouse Monoclonal Antibody [clone: 7H150]

Lieferant: United States Biological
USBIC2549-18EA 520 EUR
USBIC2549-18
Anti-CD270 Mouse Monoclonal Antibody [clone: 7H150]
Antikörper
Type: Primary
Antigen: CD270
Clonality: Monoclonal
Clone: 7H150
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR