Anti-BCAM Mouse monoclonal antibody [clone: BRIC221]

Lieferant: United States Biological
USBIC2548-98REA 835 EUR
USBIC2548-98R
Anti-BCAM Mouse monoclonal antibody [clone: BRIC221]
Antikörper
Recommended dilution: Flow Cytometry:

Type: Primary
Antigen: BCAM (basal cell adhesion molecule (Lutheran blood group))
Clonality: Monoclonal
Clone: BRIC221
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR