Anti-CD200R1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 11C4]

Lieferant: United States Biological
USBIC2548-96W8EA 710 EUR
USBIC2548-96W8
Anti-CD200R1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 11C4]
Antikörper
Do not freeze!

Type: Primary
Antigen: CD200R1 (CD200 receptor 1)
Clonality: Monoclonal
Clone: 11C4
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR