Anti-CD200R1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 6A389]

Lieferant: United States Biological
USBIC2548-96W4EA 536 EUR
USBIC2548-96W4
Anti-CD200R1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 6A389]
Antikörper
Type: Primary
Antigen: CD200R1 (CD200 receptor 1)
Clonality: Monoclonal
Clone: 6A389
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR