Anti-CD200 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: OX-104]

Lieferant: United States Biological
USBIC2548-96B1EA 746 EUR
USBIC2548-96B1
Anti-CD200 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: OX-104]
Antikörper
Type: Primary
Antigen: CD200 (CD200 molecule)
Clonality: Monoclonal
Clone: OX-104
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR