Anti-ALCAM Mouse Monoclonal Antibody [clone: 3F8B12]

Lieferant: United States Biological
USBIC2548-90P3EA 0 EUR
USBIC2548-90P3
Anti-ALCAM Mouse Monoclonal Antibody [clone: 3F8B12]
Antikörper
Recommended dilution: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunochemistry: 1:200-1:400

Type: Primary
Antigen: ALCAM (activated leukocyte cell adhesion molecule)
Clonality: Monoclonal
Clone: 3F8B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR