Anti-CD165 Mouse Monoclonal Antibody [clone: 7H142]

Lieferant: United States Biological
USBIC2548-90KDEA 370 EUR
USBIC2548-90KD
Anti-CD165 Mouse Monoclonal Antibody [clone: 7H142]
Antikörper
Type: Primary
Antigen: CD165
Clonality: Monoclonal
Clone: 7H142
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR