Anti-CD160 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 9i94]

Lieferant: United States Biological
USBIC2548-88ACEA 487 EUR
USBIC2548-88AC
Anti-CD160 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 9i94]
Antikörper
Recommended dilution: Flow Cytometry:

Type: Primary
Antigen: CD160 (CD160 molecule)
Clonality: Monoclonal
Clone: 9i94
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR