Anti-CD160 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 12K49]
Lieferant: United States Biological
USBIC2548-87TEA 654 EUR
USBIC2548-87T
Anti-CD160 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 12K49]
Antikörper
Do not freeze!
Type: Primary
Antigen: CD160 (CD160 molecule)
Clonality: Monoclonal
Clone: 12K49
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human