Anti-CD158b Mouse Monoclonal Antibody [clone: 6A379]

Lieferant: United States Biological
USBIC2548-87CEA 764 EUR
USBIC2548-87C
Anti-CD158b Mouse Monoclonal Antibody [clone: 6A379]
Antikörper
Recommended dilution: Flow Cytometry: 1:10

Type: Primary
Antigen: CD158b
Clonality: Monoclonal
Clone: 6A379
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR