Anti-TNFSF8 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: RM153]

Lieferant: United States Biological
USBIC2548-84J7EA 329 EUR
USBIC2548-84J7
Anti-TNFSF8 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: RM153]
Antikörper
Type: Primary
Antigen: TNFSF8 (tumor necrosis factor (ligand) superfamily, member 8)
Clonality: Monoclonal
Clone: RM153
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR