Anti-SLAMF1 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIC2548-81Y1EA 587 EUR
USBIC2548-81Y1
Anti-SLAMF1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000 Dot Blot: 1:500

Type: Primary
Antigen: SLAMF1 (signaling lymphocytic activation molecule family member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR