Anti-SLAMF1 Rat Monoclonal Antibody (Biotin) [clone: 10B773]
Lieferant: United States Biological
USBIC2548-81U6EA 301 EUR
USBIC2548-81U6
Anti-SLAMF1 Rat Monoclonal Antibody (Biotin) [clone: 10B773]
Antikörper
Recommended dilution: Flow Cytometry:1:10;
Type: Primary
Antigen: SLAMF1 (signaling lymphocytic activation molecule family member 1)
Clonality: Monoclonal
Clone: 10B773
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse