Anti-CDw150 Mouse monoclonal antibody [clone: 9H94]

Lieferant: United States Biological
USBIC2548-81TEA 602 EUR
USBIC2548-81T
Anti-CDw150 Mouse monoclonal antibody [clone: 9H94]
Antikörper
Recommended dilution: Western Blot: 2?g/ml

Type: Primary
Antigen: CDw150
Clonality: Monoclonal
Clone: 9H94
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR