Anti-BSG Mouse Monoclonal Antibody [clone: UM-8D6]

Lieferant: United States Biological
USBIC2548-80V1EA 578 EUR
USBIC2548-80V1
Anti-BSG Mouse Monoclonal Antibody [clone: UM-8D6]
Antikörper
Recommended dilution: Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: BSG (basigin (Ok blood group))
Clonality: Monoclonal
Clone: UM-8D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR