Anti-CD137 Ligand Mouse Monoclonal Antibody [clone: 6D296]

Lieferant: United States Biological
USBIC2548-55EA 434 EUR
USBIC2548-55
Anti-CD137 Ligand Mouse Monoclonal Antibody [clone: 6D296]
Antikörper
Recommended dilution: Flow Cytometry:1:1000; Western Blot: 1:2000

Type: Primary
Antigen: CD137 Ligand
Clonality: Monoclonal
Clone: 6D296
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR