Anti-TNFRSF9 Mouse Monoclonal Antibody [clone: 4B4-1]

Lieferant: United States Biological
USBIC2548-50HEA 321 EUR
USBIC2548-50H
Anti-TNFRSF9 Mouse Monoclonal Antibody [clone: 4B4-1]
Antikörper
Type: Primary
Antigen: TNFRSF9 (tumor necrosis factor receptor superfamily, member 9)
Clonality: Monoclonal
Clone: 4B4-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR