Anti-IL3RA Rat Monoclonal Antibody [clone: 6A367]

Lieferant: United States Biological
USBIC2462-04EA 481 EUR
USBIC2462-04
Anti-IL3RA Rat Monoclonal Antibody [clone: 6A367]
Antikörper
Type: Primary
Antigen: IL3RA (interleukin 3 receptor, alpha (low affinity))
Clonality: Monoclonal
Clone: 6A367
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR