Anti-VCAM1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: MVCAM A (429)]

Lieferant: United States Biological
USBIC2447-02PEA 553 EUR
USBIC2447-02P
Anti-VCAM1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: MVCAM A (429)]
Antikörper
Recommended dilution: Flow Cytometry:1:10;
Do not freeze!

Type: Primary
Antigen: VCAM1 (vascular cell adhesion molecule 1)
Clonality: Monoclonal
Clone: MVCAM A (429)
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR