Anti-VCAM1 Rat Monoclonal Antibody [clone: 5K220]

Lieferant: United States Biological
USBIC2446-79AXEA 407 EUR
USBIC2446-79AX
Anti-VCAM1 Rat Monoclonal Antibody [clone: 5K220]
Antikörper
Type: Primary
Antigen: VCAM1 (vascular cell adhesion molecule 1)
Clonality: Monoclonal
Clone: 5K220
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1 Kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR