Anti-VCAM1 Mouse Monoclonal Antibody [clone: 6A518 (1G11B1)]

Lieferant: United States Biological
USBIC2446-77AEA 515 EUR
USBIC2446-77A
Anti-VCAM1 Mouse Monoclonal Antibody [clone: 6A518 (1G11B1)]
Antikörper
Recommended dilution: Flow Cytometry: 1:10

Type: Primary
Antigen: VCAM1 (vascular cell adhesion molecule 1)
Clonality: Monoclonal
Clone: 6A518 (1G11B1)
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR