Anti-CD99 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIC2444-02BEA 559 EUR
USBIC2444-02B
Anti-CD99 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: CD99 (CD99 molecule)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR