Anti-CD99 Mouse Monoclonal Antibody [clone: MEM-131]

Lieferant: United States Biological
USBIC2444-02AEA 538 EUR
USBIC2444-02A
Anti-CD99 Mouse Monoclonal Antibody [clone: MEM-131]
Antikörper
Type: Primary
Antigen: CD99 (CD99 molecule)
Clonality: Monoclonal
Clone: MEM-131
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR