Anti-CD98 Mouse Monoclonal Antibody [clone: MEM-108]

Lieferant: United States Biological
USBIC2443-86QEA 578 EUR
USBIC2443-86Q
Anti-CD98 Mouse Monoclonal Antibody [clone: MEM-108]
Antikörper
Recommended dilution: Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: CD98
Clonality: Monoclonal
Clone: MEM-108
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR