Anti-C5AR1 Mouse monoclonal antibody [clone: P12/1]

Lieferant: United States Biological
USBIC2439-61H1EA 578 EUR
USBIC2439-61H1
Anti-C5AR1 Mouse monoclonal antibody [clone: P12/1]
Antikörper
Recommended dilution: Immunochemistry: 1:200; Flow Cytometry: 1:100-1:50

Type: Primary
Antigen: C5AR1 (complement component 5a receptor 1)
Clonality: Monoclonal
Clone: P12/1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR