Anti-C5AR1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 10B2112]

Lieferant: United States Biological
USBIC2439-61C5EA 0 EUR
USBIC2439-61C5
Anti-C5AR1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 10B2112]
Antikörper
Recommended dilution: Flow Cytometry:1:10;
Do not freeze!

Type: Primary
Antigen: C5AR1 (complement component 5a receptor 1)
Clonality: Monoclonal
Clone: 10B2112
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR