Anti-C5AR1 Mouse monoclonal antibody [clone: 3H1738]

Lieferant: United States Biological
USBIC2439-60D-500EA 634 EUR
USBIC2439-60D-500 USBIC2439-60D-1
Anti-C5AR1 Mouse monoclonal antibody [clone: 3H1738]
Antikörper
Type: Primary
Antigen: C5AR1 (complement component 5a receptor 1)
Clonality: Monoclonal
Clone: 3H1738
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR