Anti-CD86 Rat Monoclonal Antibody [clone: PO3]

Lieferant: United States Biological
USBIC2438-05UEA 437 EUR
USBIC2438-05U
Anti-CD86 Rat Monoclonal Antibody [clone: PO3]
Antikörper
Recommended dilution: Flow Cytometry:

Type: Primary
Antigen: CD86 (CD86 molecule)
Clonality: Monoclonal
Clone: PO3
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR