Anti-CD86 Mouse Monoclonal Antibody [clone: 10B2092]
Lieferant: United States Biological
USBIC2438-05REA 552 EUR
USBIC2438-05R
Anti-CD86 Mouse Monoclonal Antibody [clone: 10B2092]
Antikörper
Recommended dilution: Flow Cytometry: 1:40;
Type: Primary
Antigen: CD86 (CD86 molecule)
Clonality: Monoclonal
Clone: 10B2092
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken