Anti-CD83 Mouse Monoclonal Antibody [clone: 7H190]

Lieferant: United States Biological
USBIC2435-04XEA 473 EUR
USBIC2435-04X
Anti-CD83 Mouse Monoclonal Antibody [clone: 7H190]
Antikörper
Type: Primary
Antigen: CD83 (CD83 molecule)
Clonality: Monoclonal
Clone: 7H190
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR