Anti-CD81 Hamster Monoclonal Antibody (Biotin) [clone: 10B2068]

Lieferant: United States Biological
USBIC2433-10L2EA 304 EUR
USBIC2433-10L2
Anti-CD81 Hamster Monoclonal Antibody (Biotin) [clone: 10B2068]
Antikörper
Type: Primary
Antigen: CD81 (CD81 molecule)
Clonality: Monoclonal
Clone: 10B2068
Conjugation: Biotin
Epitope:
Host: Hamster
Isotype: IgG1
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR