Anti-CD81 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 7H100]

Lieferant: United States Biological
USBIC2433-10CEA 414 EUR
USBIC2433-10C
Anti-CD81 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 7H100]
Antikörper
Recommended dilution: Flow Cytometry: 1:50

Type: Primary
Antigen: CD81 (CD81 molecule)
Clonality: Monoclonal
Clone: 7H100
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR