Anti-CD81 Mouse Monoclonal Antibody [clone: 7H100]

Lieferant: United States Biological
USBIC2433-10AEA 370 EUR
USBIC2433-10A
Anti-CD81 Mouse Monoclonal Antibody [clone: 7H100]
Antikörper
Type: Primary
Antigen: CD81 (CD81 molecule)
Clonality: Monoclonal
Clone: 7H100
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR