Anti-CD80 Rat Monoclonal Antibody [clone: 5K18]

Lieferant: United States Biological
USBIC2432-14XEA 407 EUR
USBIC2432-14X
Anti-CD80 Rat Monoclonal Antibody [clone: 5K18]
Antikörper
Type: Primary
Antigen: CD80 (CD80 molecule)
Clonality: Monoclonal
Clone: 5K18
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR