Anti-CD80 Rabbit Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBIC2432-03C-50EA 425 EUR
USBIC2432-03C-50
Anti-CD80 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: CD80 (CD80 molecule)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR