Anti-CD80 Mouse Monoclonal Antibody (Biotin) [clone: 7H211]
Lieferant: United States Biological
USBIC2432-01FEA 675 EUR
USBIC2432-01F
Anti-CD80 Mouse Monoclonal Antibody (Biotin) [clone: 7H211]
Antikörper
Type: Primary
Antigen: CD80 (CD80 molecule)
Clonality: Monoclonal
Clone: 7H211
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human