Anti-CD79A Mouse Monoclonal Antibody (Biotin) [clone: 10B1965]

Lieferant: United States Biological
USBIC2431-14EEA 414 EUR
USBIC2431-14E
Anti-CD79A Mouse Monoclonal Antibody (Biotin) [clone: 10B1965]
Antikörper
Type: Primary
Antigen: CD79A (CD79a molecule, immunoglobulin-associated alpha)
Clonality: Monoclonal
Clone: 10B1965
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR