Anti-CD79A Mouse Monoclonal Antibody [clone: 10B1965]

Lieferant: United States Biological
USBIC2431-14BEA 645 EUR
USBIC2431-14B
Anti-CD79A Mouse Monoclonal Antibody [clone: 10B1965]
Antikörper
Recommended dilution: Flow Cytometry: 10 ?g/ml; Immunoprecipitation: 20 ?g/ml

Type: Primary
Antigen: CD79A (CD79a molecule, immunoglobulin-associated alpha)
Clonality: Monoclonal
Clone: 10B1965
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR