Anti-CD79B Hamster Monoclonal Antibody (Biotin) [clone: 5K35]

Lieferant: United States Biological
USBIC2431-06CEA 462 EUR
USBIC2431-06C
Anti-CD79B Hamster Monoclonal Antibody (Biotin) [clone: 5K35]
Antikörper
Type: Primary
Antigen: CD79B (CD79b molecule, immunoglobulin-associated beta)
Clonality: Monoclonal
Clone: 5K35
Conjugation: Biotin
Epitope:
Host: Hamster
Isotype: IgG Kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR