Anti-CD79B Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 7H118]

Lieferant: United States Biological
USBIC2431-05JEA 524 EUR
USBIC2431-05J
Anti-CD79B Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 7H118]
Antikörper
Type: Primary
Antigen: CD79B (CD79b molecule, immunoglobulin-associated beta)
Clonality: Monoclonal
Clone: 7H118
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR