Anti-CD72 Mouse Monoclonal Antibody [clone: BL-A/A11]

Lieferant: United States Biological
USBIC2428-41YEA 421 EUR
USBIC2428-41Y
Anti-CD72 Mouse Monoclonal Antibody [clone: BL-A/A11]
Antikörper
Type: Primary
Antigen: CD72 (CD72 molecule)
Clonality: Monoclonal
Clone: BL-A/A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR